Orders LinkContactUsLinkComponent
EasySep™ Mouse Memory CD4+ T Cell Isolation Kits
EasySep™ Mouse Memory CD4+ T Cell Isolation Kits
EasySep™ mouse memory CD4+ T cell isolation kit, 1×10⁹ cells
Katalog # STMC19767
Marketsource
Leverandør:   Stemcell Technologies
EasySep™ Mouse Memory CD4+ T Cell Isolation Kits
Katalog # STMC19767
Leverandør:  Stemcell Technologies
Marketsource
EasySep™ Mouse Memory CD4+ T Cell Isolation Kits
EasySep™ Mouse Memory CD4+ T Cell Isolation Kits
EasySep™ mouse memory CD4+ T cell isolation kit, 1×10⁹ cells
Price
5.670,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Immunomagnetic isolation of untouched mouse memory CD4 T cells
  • Fast, easy-to-use and column-free
  • Up to 96% purity
  • Untouched, viable cells
The EasySep™ Mouse Memory CD4+ T Cell Isolation Kit is designed to isolate pan-memory CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-memory CD4+ T cells , featuring RapidSphere™ technology.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
  • Beskrivelse:
    EasySep™ mouse memory CD4+ T cell isolation kit, 1×10⁹ cells
  • Seq:
    01