0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
PBS BUFFER 2000ML/POUCH 1 * 1 Each
BSBTAR0030
:
Boster Bio
PBS BUFFER 2000ML/POUCH 1 * 1 Each
BSBTAR0030
:
Boster Bio
About Boster
AR0030
Literature reference - PubMed ID 17487418
Literature reference - PubMed ID 20037813
Literature reference - PubMed ID 20402666
Literature reference - PubMed ID 20673188
Literature reference - PubMed ID 20863781
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Ansigtsservietter
: VWR Collection
VWR®, Nitril handsker, NITRILE
: VWR Collection
pH-teststrimler, MQuant®, Supelco®
: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Vand, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Handsker, nitril, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Serologiske pipetter, Standard type
: VWR Collection