Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Conductivity standard solution, 147 µS/cm, 250 mL
Katalog # HACHLZW9700.99
Leverandør:   Hach
Conductivity standards
Katalog # HACHLZW9700.99
Leverandør:  Hach
CAS nummer:  
Conductivity standards
Conductivity standards
Conductivity standard solution, 147 µS/cm, 250 mL
CAS nummer:  
Price
217,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specifikationer
Specifikationer
  • Pakn:
    250 mL
  • Beskrivelse:
    Conductivity standard solution
  • Konduktivitet:
    147 µS/cm