Trays
Flask tray with threaded holes to attach 25 to 1000 ml SC flask clamps, SH plate holders and SR racks, for LSB12
Katalog # 462-0537
Leverandør: GRANT INSTRUMENTS
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Flask tray with threaded holes to attach 25 to 1000 ml SC flask clamps, SH plate holders and SR racks, for LSB12
- Seq:01