Orders LinkContactUsLinkComponent
Digital dry block heaters
Digital dry block heaters
Dry Block Heater 4, 4, 412 W, RT*...120 °C, ±0,2 °C, 151×456×73 mm, 2 kg
Katalog # 460-0263
Digital dry block heaters
Katalog # 460-0263
Leverandør:  
Digital dry block heaters
Digital dry block heaters
Dry Block Heater 4, 4, 412 W, RT*...120 °C, ±0,2 °C, 151×456×73 mm, 2 kg
Discontinued
Product Details & Documents
Digital block heaters which hold from one up to four aluminum blocks, provide precise temperature control in small vessels. The PT1000 temperature sensor allows temperature control directly in the sample vessel. It is used with PCR tubes, PCR strips, Greiner tubes, microplates and cuvettes. The block heaters are ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood/urea nitrogen determinations and in situ hybridisations.
  • Stepless adjustable temperature from ambient up to 120 °C
  • Timer: Countdown, adjustable from 1 min to 99h 59 min
  • Counter: Shows heating time
  • Highly versatile with a large number of aluminium alloy blocks
  • Fixed safety circuit and error code display
  • Acoustic signal at end of test
Delivery information: Supplied with PT1000 temperature sensor, but without heating blocks, blocks must be ordered separately.
Caution: * RT = Rumtemperatur
Specifikationer
  • Opvarmnings kapacitet:
    412 W
  • Antal blokke:
    4
  • Temperaturområde:
    RT*...120 °C
  • Vægt:
    2 kg
  • Seq:
    07
  • B×D×H:
    151×456×73 mm
  • Temp. stabilitet:
    ±0,2 °C
  • Model:
    Dry Block Heater 4
Accessories
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 20×11,5 mm Ø holes
    Single block, 20×11,5 mm Ø holes
    460-1009
    Eppendorf Tubes®, 1,5 ml
    36,9 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Model
    Område
    Tilgængelighed
    Price
  • Temperature sensor, coated (SafeCoat), DIN 45322
    Temperature sensor, coated (SafeCoat), DIN 45322
    460-1041
    PT 1000.90
    −10...+260 °C
    Log ind for tilgængelighed
    1 * 1 Items - 2.860,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 80× 6,4 mm Ø strips
    Single block, 80× 6,4 mm Ø strips
    460-1036
    PCR tube strips (10×8 strips)
    15,5 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 30×7,9 mm Ø holes
    Single block, 30×7,9 mm Ø holes
    460-1008
    PCR tubes, 0,5 ml
    27,6 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 4×35 mm Ø holes
    Single block, 4×35 mm Ø holes
    460-1025
    Tubes, 35 mm, with round bottom
    47,6 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 12×17,5 mm Ø holes
    Single block, 12×17,5 mm Ø holes
    460-1022
    Tubes, 15 - 16 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Double block
    Double block
    460-1038
    96-well PCR plate (1)
    15,5 mm
    Log ind for tilgængelighed
    1 * 1 Items - 2.700,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 30×8,3 mm Ø holes
    Single block, 30×8,3 mm Ø holes
    460-1018
    Tubes, 6 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 8×23,8 mm Ø holes
    Single block, 8×23,8 mm Ø holes
    460-1032
    Vials, 23 mm
    45 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 16×13,9 mm Ø holes
    Single block, 16×13,9 mm Ø holes
    460-1020
    Tubes, 12 - 13 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Støttestav, rustfrit stål, længde: 450 mm, Ø 10 mm, til alle omrørere med M10-gevind
    Støttestav, rustfrit stål, længde: 450 mm, Ø 10 mm, til alle omrørere med M10-gevind
    442-2041
    Log ind for tilgængelighed
    1 * 1 Items - 707,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 12×17,8 mm Ø holes
    Single block, 12×17,8 mm Ø holes
    460-1029
    Vials, 17 mm
    45 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 5×29 mm Ø holes
    Single block, 5×29 mm Ø holes
    460-1014
    Conical tubes, 50 ml
    47,6 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Double block
    Double block
    460-1040
    96- or 384-well plate (1)
    13,5 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.710,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, combination: 4/3/2×1,5/15/50 ml
    Single block, combination: 4/3/2×1,5/15/50 ml
    460-1016
    Centrifuge tubes, 1,5 ml/15 ml/50 ml
    39,1/44,5/47,6 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 20×15,8 mm Ø holes
    Single block, 20×15,8 mm Ø holes
    460-1028
    Vials, 15 mm
    35 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 6×28,8 mm Ø holes
    Single block, 6×28,8 mm Ø holes
    460-1034
    Vials, 28 mm
    45 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 8×25,8 mm Ø holes
    Single block, 8×25,8 mm Ø holes
    460-1033
    Vials, 25 mm
    45 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 20×13,9 mm Ø holes
    Single block, 20×13,9 mm Ø holes
    460-1021
    Tubes, 12 - 13 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 24×10,7 mm Ø holes
    Single block, 24×10,7 mm Ø holes
    460-1019
    Tubes, 10 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 20×10,9 mm Ø holes
    Single block, 20×10,9 mm Ø holes
    460-1012
    Corning tubes, 2,0 ml
    38,1 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 20×11,5 mm Ø holes
    Single block, 20×11,5 mm Ø holes
    460-1011
    VWR/Eppendorf Tubes®, 2,0 ml
    38,1 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 2×6 cuvettes 12,5 mm Ø
    Single block, 2×6 cuvettes 12,5 mm Ø
    460-1039
    Cuvettes
    25,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 8×21 mm Ø holes
    Single block, 8×21 mm Ø holes
    460-1023
    Tubes, 20 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Dobbeltmuffe
    Dobbeltmuffe
    442-2043
    Log ind for tilgængelighed
    1 * 1 Items - 336,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 12×19,1 mm Ø holes
    Single block, 12×19,1 mm Ø holes
    460-1026
    Tubes, 17 - 18 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, combination: 6/10/5×0,5/1,5/2 ml
    Single block, combination: 6/10/5×0,5/1,5/2 ml
    460-1017
    Microtubes, 0,5 ml/1,5 ml/2,0 ml
    27,6/39,1/38,1 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 12×19,7 mm Ø holes
    Single block, 12×19,7 mm Ø holes
    460-1030
    Vials, 19 mm
    45 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 20×12,7 mm Ø holes
    Single block, 20×12,7 mm Ø holes
    460-1027
    Vials, 12 mm
    30 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 12×17,1 mm Ø holes
    Single block, 12×17,1 mm Ø holes
    460-1013
    Conical tubes, 15 ml
    44,5 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 6×26,2 mm Ø holes
    Single block, 6×26,2 mm Ø holes
    460-1024
    Tubes, 25 mm, with round bottom
    48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 64× 6,4 mm Ø holes
    Single block, 64× 6,4 mm Ø holes
    460-1037
    PCR tubes (64)
    20,2 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Holder til ETS-D5 or VT-5. til stativstang
    Holder til ETS-D5 or VT-5. til stativstang
    431-2888
    Log ind for tilgængelighed
    1 * 1 Items - 312,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 9×21,7 mm Ø holes
    Single block, 9×21,7 mm Ø holes
    460-1031
    Vials, 21 mm
    45 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, combination: 6/5/3×6/12/13/25 mm Ø holes
    Single block, combination: 6/5/3×6/12/13/25 mm Ø holes
    460-1015
    Test tubes, 6 ml/12 - 13 ml/25 ml
    48,4/48,4/48,4 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 20×11,1 mm Ø holes
    Single block, 20×11,1 mm Ø holes
    460-1010
    Microtubes 1,5 ml
    39,1 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.120,00 kr
Produktinformation
  • Katalog #
    Til
    Dybde
    Tilgængelighed
    Price
  • Single block, 15×16,4 mm Ø holes
    Single block, 15×16,4 mm Ø holes
    460-1035
    Vials, 16 mm
    45 mm
    Log ind for tilgængelighed
    1 * 1 Items - 1.180,00 kr
Produktinformation
  • Katalog #
    Model
    Område
    Tilgængelighed
    Price
  • Stainless steel temperature sensor, DIN 45322
    Stainless steel temperature sensor, DIN 45322
    442-0584
    PT 1000.60
    −20...+350 °C
    Log ind for tilgængelighed
    1 * 1 Items - 1.350,00 kr