Orders LinkContactUsLinkComponent
Dropping pipettes
Dropping pipettes
Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15
Katalog # BIBBSBD050
Leverandør:   SciLabware
Dropping pipettes
Katalog # BIBBSBD050
Leverandør:  SciLabware
Dropping pipettes
Dropping pipettes
Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15
Price
862,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Soda-lime glass.
  • With integral ground glass cone on pipette
  • Supplied with rubber teats
  • Bottles available separately
Specifikationer
  • Beskrivelse:
    Dropping pipettes with silicone teat, for 50 ml bottles, joint size 14/15
  • Seq:
    01