Orders LinkContactUsLinkComponent
VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan PH objective, 20×/0,40
Catalog # 630-2166
Supplier:   VWR Collection
VWR®, IOS Objectives for Inverted Microscopes
Catalog # 630-2166
Supplier:  VWR Collection
VWR®, IOS Objectives for Inverted Microscopes
VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan PH objective, 20×/0,40
Price
4.470,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These objectives are designed to be used with inverted microscopes.
  • High resolution and contrast
  • Ensure field flatness up to F.N. 22, based on infinity-corrected optical system
  • Long Working Distance provides a wider working space between the lens surface and the object
Ideal for the observation of culture specimens, clinical examinations and cell testing.
Specifications
  • Forstørrelse:
    20×/0,40
  • Beskrivelse:
    IOS LWD W-Plan PH objective
  • Seq:
    205