Orders LinkContactUsLinkComponent
Accessories for storage cabinets
Second level drawer realised as perforated insert, stainless steel, max. load 25 kg, 715×415×37 mm
Katalog # 135-1937
Leverandør:   asecos
Accessories for storage cabinets
Katalog # 135-1937
Leverandør:  asecos
Accessories for storage cabinets
Accessories for storage cabinets
Second level drawer realised as perforated insert, stainless steel, max. load 25 kg, 715×415×37 mm
Price
2.370,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Second level drawer realised as perforated insert, stainless steel, max. load 25 kg, 715×415×37 mm
  • Seq:
    28