Accessories for storage cabinets
Second level drawer realised as perforated insert, stainless steel, max. load 25 kg, 715×415×37 mm
Katalog # 135-1937
Leverandør: asecos
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Second level drawer realised as perforated insert, stainless steel, max. load 25 kg, 715×415×37 mm
- Seq:28