Orders LinkContactUsLinkComponent
Cleanroom mops, PocketMop™
Cleanroom mops, PocketMop™
PocketMop™ blue microfibre head mop, 356 mm
Katalog # MICRPKMAA-14
Leverandør:   MICRONOVA
Cleanroom mops, PocketMop™
Katalog # MICRPKMAA-14
Leverandør:  MICRONOVA
Cleanroom mops, PocketMop™
Cleanroom mops, PocketMop™
PocketMop™ blue microfibre head mop, 356 mm
Price
601,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents
These mop can be used for controlled environments and is economical enough to compete with commercial mops that are often used in support or non-classified areas.
  • Lightweight and easy to maneuver
  • 100% smooth knit polyester
  • Compatible with strong disinfectants
The swivel joint rotates 360° but can be locked in a 180° movement for more precise, controlled cleaning. Economical enough for single use but can be laundered and reused.
Specifikationer
  • Beskrivelse:
    PocketMop™ blue microfibre head mop, 356 mm
  • Seq:
    6