Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cleanroom mops, PocketMop™

PocketMop™ NovaPoly™ flat mop head, 356 mm
  MICRPKM11-14
 :   MICRONOVA
Cleanroom mops, PocketMop™
  MICRPKM11-14
 :  MICRONOVA
Cleanroom mops, PocketMop™
Cleanroom mops, PocketMop™

PocketMop™ NovaPoly™ flat mop head, 356 mm

 
601,00 kr
   
1 * 4 Items
 

These mop can be used for controlled environments and is economical enough to compete with commercial mops that are often used in support or non-classified areas.

  • Lightweight and easy to maneuver
  • 100% smooth knit polyester
  • Compatible with strong disinfectants

The swivel joint rotates 360° but can be locked in a 180° movement for more precise, controlled cleaning. Economical enough for single use but can be laundered and reused.

 
  • Beskrivelse:
    PocketMop™ NovaPoly™ flat mop head, 356 mm
  • Seq:
    5