VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan PH objective, 10×/0,25
630-2165
These objectives are designed to be used with inverted microscopes.
- High resolution and contrast
- Ensure field flatness up to F.N. 22, based on infinity-corrected optical system
- Long Working Distance provides a wider working space between the lens surface and the object
Ideal for the observation of culture specimens, clinical examinations and cell testing.
- Forstørrelse:10×/0,25
- Beskrivelse:IOS LWD W-Plan PH objective
- Seq:204