Orders LinkContactUsLinkComponent
VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan PH objective, 10×/0,25
  630-2165
VWR®, IOS Objectives for Inverted Microscopes
  630-2165
VWR®, IOS Objectives for Inverted Microscopes
VWR®, IOS Objectives for Inverted Microscopes
IOS LWD W-Plan PH objective, 10×/0,25
3.130,00 kr
1 * 1 Items
These objectives are designed to be used with inverted microscopes.
  • High resolution and contrast
  • Ensure field flatness up to F.N. 22, based on infinity-corrected optical system
  • Long Working Distance provides a wider working space between the lens surface and the object
Ideal for the observation of culture specimens, clinical examinations and cell testing.
  • Forstørrelse:
    10×/0,25
  • Beskrivelse:
    IOS LWD W-Plan PH objective
  • Seq:
    204