Caps, closed top
With bonded PTFE Liner, bulk, 38 - 400 mm
Katalog # EPSCB167-3800BL
Leverandør: EP SCIENTIFIC
Gå til:
Product Details & Documents
PP, white.
- Very good chemical resistance
- Very good temperature resistance
Unprocessed caps are ideal for environmental sample bottles, jars, jugs and vials.
Certifications: Manufactured under an ISO certified quality management system
Specifikationer
- Cap størrelse:38 - 400 mm
- Beskrivelse:With bonded PTFE Liner, bulk
- Seq:04
- Cat. No.:EPSCB167-3800BL
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning