Septa caps
Open top cap, without septum, 24-414
Katalog # EPSC297-2414
Leverandør: EP SCIENTIFIC
Gå til:
Product Details & Documents
PP, white.
Septa caps fit VOA vials, bottles and jars. The septum is permanently bonded to the cap and processed to meet EPA performance based specifications for volatile organic analysis.
Specifikationer
- Gevind:24-414
- Beskrivelse:Open top cap, without septum
- Seq:05
- Cat. No.:EPSC297-2414
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning
Accessories
Produktinformation
- Katalog #ModelTilØTilgængelighedPrice
- EPSC288-0022Silicone, PTFE-lined24-214 VOA vials24 mmLog ind for tilgængelighed1 * 24 Items - 483,00 kr
Produktinformation
- Katalog #ModelTilØTilgængelighedPrice
- EPSC288-7222Silicone, PTFE-lined24-214 VOA vials24 mmLog ind for tilgængelighed1 * 72 Items - 1.180,00 kr
Produktinformation
- Katalog #ModelTilØTilgængelighedPrice
- EPSC288-0022BPSilicone, PTFE-lined24-214 VOA vials24 mmLog ind for tilgængelighed1 * 2.000 Items - 18.070,00 kr
Produktinformation
- Katalog #ModelTilØTilgængelighedPrice
- EPSCS24-400-S2Silicone, PTFE-lined24-214 VOA vials24 mmLog ind for tilgængelighed1 * 144 Items - 2.140,00 kr