Screw caps
Caps, PP, white, LDPE foam lined, 24-410
Catalog # EPSC067-2410WBP
Supplier: EP SCIENTIFIC
Jump to:
Product Details & Documents
LDPE-lined white caps.
- For general-purpose HDPE bulk separate containers
- Same high quality caps used on certified HDPE environmental sample containers
Ordering information: Bottles sold separately (IR013-series and IR012-series).
Specifications
- Gevind:24-410
- Beskrivelse:Caps, PP, white, LDPE foam lined
- Seq:01
- Cat. No.:EPSC067-2410WBP
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning