Spray gun
Recirculating tee fitting (9,5 mm Flaretek® tube fitting × 9,5 mm Flaretek® tube fitting × 3,2 mm compression)
Katalog # ENTE215-138
Leverandør: ENTEGRIS
Gå til:
Product Details & Documents
High purity PFA (all wetted parts).
- With 4,8 mm orifice and ³/₈" Flaretek® tube fitting or ¹/₂" compression connections
- Max. pressure 5,52 bar
Used in fume hoods and wet processing stations to help protect against corrosion.
Specifikationer
- Beskrivelse:Recirculating tee fitting (9,5 mm Flaretek® tube fitting × 9,5 mm Flaretek® tube fitting × 3,2 mm compression)
- Seq:07
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Repair tool to access front and rear diaphragm of gunRepair tool to access front and rear diaphragm of gunENTE213-80Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spoolRepair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spoolENTE421-06Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement cap, natural
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement nozzle
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement cap, orange
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement cap, blue



