Eddikesyre glacial ≥98%, TECHNICAL
Plastic drum, 25 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
- CAS Number:64-19-7
- Formula:H₃CCOOH
- MDL Number:MFCD00036152
- Molecular Weight:60.05 g/mol
- Boiling Pt:118 °C (1013 hPa)
- Melting Pt:17 °C
- Density:1,05 g/cm³ (20 °C)
- Flash Pt:38,5 °C
- Storage Temperature:Stue temperatur
- EINECS:200-580-7
- UN:2789
- ADR:8 (3),II
- REACH:01-2119475328-30
Specifications
Specification Test Results
- Assay:Min. 98 %
Specifications
- Pakning:Plastic drum
- Pakn:25 L



