Orders LinkContactUsLinkComponent
Caps for tubes
Caps for tubes
Cap for 12 and 13 mm tubes, blå
Katalog # DELT307105
Leverandør:   DELTALAB
Caps for tubes
Katalog # DELT307105
Leverandør:  DELTALAB
Caps for tubes
Cap for 12 and 13 mm tubes, blå
Price
441,00 kr
Log ind for tilgængelighed
1 * 2.000 Items
Product Details & Documents
Polyethylene caps for all types of polypropylene and polystyrene tubes.
Specifikationer
  • Farve:
    blå
  • Beskrivelse:
    Cap for 12 and 13 mm tubes
  • Seq:
    08