Hand towels, SCOTT® PERFORMANCE
SCOTT® Performance, 1 ply, small, hvid, 210×215 mm, 15×300 towels
KIMW6659
These 1-ply interfolded hand towels are made from strong, highly absorbent AIRFLEX* fabric and supplied in cost-saving ‘compressed’ packs (with 18% more towels per case).
- Soft yet highly absorbent, even when wet
- Dry hands hygienically and efficiently
Ideal for busy office and hotel washrooms.
- Pakket:15×300 towels
- Farve:hvid
- Beskrivelse:SCOTT® Performance, 1 ply, small
- L×B:210×215 mm
Complementary Products
- BeskrivelseB×D×H
- 115-1780Dispenser for C-fold hand towels, AQUARIUS* 6945, white ABS, with window, large280×146×412 mm1 * 1 Items - 415,00 kr
- 115-1783Dispenser for interfolded hand towels, KIMBERLY-CLARK PROFESSIONAL*, 8971, stainless steel237×118×349 mm1 * 1 Items - 2.850,00 kr
- 115-1782Dispenser for C-fold hand towels, AQUARIUS* 6956, white ABS, with window, small307×176×155 mm1 * 1 Items - 277,00 kr