Beakers, with handle, PP
2000 ml, 206 mm, 151 mm, 20 ml
Katalog # BURK7204-2000
Leverandør: BURKLE
Gå til:
Product Details & Documents
PP, highly transparent.
- With embossed graduation printed in blue
- Ergonomic open or close handle for secure grip
- Functional spout to minimize spills
Ideal for industrial applications.
Specifikationer
- Udv.Ø:151 mm
- Inddelings-interval:20 ml
- Kapacitet:2000 ml
- Højde:206 mm
- Seq:03