Orders LinkContactUsLinkComponent
Testkits, sulfat test, til Spectroquant®, Supelco®
Testkits, sulfat test, til Spectroquant®, Supelco®
Sulphate test, photometric, 5 - 300 mg/L SO42−, Barium sulphate, turbidimetric, Move 100, Nova 60 A, Prove 100 (plus), Prove 300 (plus), Prove 600 (plus)
Katalog # 1.02537.0001
Leverandør:   Merck
Testkits, sulfat test, til Spectroquant®, Supelco®
Katalog # 1.02537.0001
Leverandør:  Merck
CAS nummer:  
Testkits, sulfat test, til Spectroquant®, Supelco®
Testkits, sulfat test, til Spectroquant®, Supelco®
Sulphate test, photometric, 5 - 300 mg/L SO42−, Barium sulphate, turbidimetric, Move 100, Nova 60 A, Prove 100 (plus), Prove 300 (plus), Prove 600 (plus)
CAS nummer:  
Price
kr1,080.00
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
Highly stable, ready to use reagent mixtures in a user friendly design.
Free selection of appropriate cell format according to the measuring range.
Long shelf life up to 3 years all at room temperature.
Automatic selection of the correct method with help of AutoSelector barcode identification.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Specifikationer
Specifikationer
  • Kemisk metode:
    Barium sulphate, turbidimetric
  • Beskrivelse:
    Sulphate test, photometric
  • Til enhed:
    Move 100, Nova 60 A, Prove 100 (plus), Prove 300 (plus), Prove 600 (plus)
  • Område:
    5 - 300 mg/L SO<sub>4</sub><sup>2−</sup>
  • Cat. No.:
    1.02537.0001
  • M-Clarity quality level:
    MQ100