Orders LinkContactUsLinkComponent
Accessories for transilluminators
Accessories for transilluminators
UV tube, 15 W, 312 nm, length 451 mm
Katalog # CONSVLT15-M
Leverandør:   CONSORT
Accessories for transilluminators
Katalog # CONSVLT15-M
Leverandør:  CONSORT
Accessories for transilluminators
Accessories for transilluminators
UV tube, 15 W, 312 nm, length 451 mm
Price
876,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    UV tube, 15 W, 312 nm, length 451 mm
  • Seq:
    10