Orders LinkContactUsLinkComponent
Accessories for transilluminators
Accessories for transilluminators
UV tube, 6 W, 365 nm, length 226 mm
Katalog # CONSVLT6-L
Leverandør:   CONSORT
Accessories for transilluminators
Katalog # CONSVLT6-L
Leverandør:  CONSORT
Accessories for transilluminators
Accessories for transilluminators
UV tube, 6 W, 365 nm, length 226 mm
Price
822,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    UV tube, 6 W, 365 nm, length 226 mm
  • Seq:
    04