Accessories for transilluminators
UV tube, 15 W, 254 nm, length 451 mm
Catalog # CONSVLT15-C
Supplier: CONSORT
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:UV tube, 15 W, 254 nm, length 451 mm
- Seq:09