Orders LinkContactUsLinkComponent
Precision hydrometers, ASTM E100
Precision hydrometers, ASTM E100
135 H-62, 1,500 - 1,550 sp/gr, 0,001 sp/gr, 260 mm
Katalog # SCNL3222021
Leverandør:   Ludwig Schneider
Precision hydrometers, ASTM E100
Katalog # SCNL3222021
Leverandør:  Ludwig Schneider
Precision hydrometers, ASTM E100
Precision hydrometers, ASTM E100
135 H-62, 1,500 - 1,550 sp/gr, 0,001 sp/gr, 260 mm
Price
258,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These precision ASTM E100 hydrometers are designed for specific gravity or relative density.
  • Reference temperature: 15,5 °C (60/60 °F)
  • Without thermometer
  • Suitable for calibration
Ordering information: Available with either a works or a DAkkS calibration certificate on request
Packaging: Each packed in a square plastic sleeve.
Specifikationer
  • Længde:
    260 mm
  • Seq:
    18
  • Inddeling:
    0,001 sp/gr
  • Model:
    135 H-62
  • Område:
    1,500 - 1,550 sp/gr
  • Cat. No.:
    SCNL3222021
  • Referencetemperatur:
    15,5 °C