Reagent containers for Microlab® STAR line
Reagent container without lid, non self-standing, Non self-standing, 50 ml, 90×20×60 mm, gennemsigtig, Individually wrapped
Katalog # HAMR187297
Marketsource
Leverandør: HAMILTON ROBOTICS
Reagent containers for Microlab® STAR line
Katalog # HAMR187297
Leverandør: HAMILTON ROBOTICS
Marketsource
Gå til:
Product Details & Documents
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
- Special bottom geometry minimises residual volumes
- With volume markings, without or with lid
- With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
- IVD compliant
- Autoclavable
Bestillingsoplysninger: The carriers are not included and need to be ordered separately.
Documents
Specifikationer
- Pakket:Individually wrapped
- Model:Non self-standing
- Kapacitet:50 ml
- Farve:gennemsigtig
- Beskrivelse:Reagent container without lid, non self-standing
- B×D×H:90×20×60 mm



