Orders LinkContactUsLinkComponent
Body lotion, Lindesa®
Body lotion, Lindesa®
Body lotion, Lindesa®, 50 ml tube
  FAWE13640-010
Body lotion, Lindesa®
  FAWE13640-010
 :  
Body lotion, Lindesa®
Body lotion, Lindesa®
Body lotion, Lindesa®, 50 ml tube
Creme Stokoderm Grip Pure 100 ml. 1 * 1 Items  115-1986
Product 115-1986 is similar to FAWE13640-010 but manufacturer and specifications may vary.
Creme Stokoderm Grip Pure 30 ml. 1 * 30 Items  115-2360
Product 115-2360 is similar to FAWE13640-010 but manufacturer and specifications may vary.
Barrier cream and care cream with beeswax is designed to care for skin exposed to minor contamination, and is also suitable for after-work care.
  • Lightly greasy, absorbs quickly
  • Special care ingredients improve the skin's elasticity and suppleness
  • Does not affect the sense of touch
  • Emulsion type: O/W, neutral pH level, silicone-free, scented
The cream strengthens the skin's immunity, prevents skin damage from exposure to external contamination, and has a soothing effect on irritated skin. It smooths and nourishes without any long-lasting, greasy film.
Caution: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
  • Beskrivelse:
    Body lotion, Lindesa®, 50 ml tube
  • Seq:
    01