Orders LinkContactUsLinkComponent
VWR®, IOS W-Plan MET Objectives
VWR®, IOS W-Plan MET Objectives
IOS W-Plan MET objectives (no cover glass), 10×/0,25 mm
Katalog # 630-2161
Leverandør:   VWR Collection
VWR®, IOS W-Plan MET Objectives
Katalog # 630-2161
Leverandør:  VWR Collection
VWR®, IOS W-Plan MET Objectives
VWR®, IOS W-Plan MET Objectives
IOS W-Plan MET objectives (no cover glass), 10×/0,25 mm
Price
1.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These objectives deliver precise performance without the need of the cover slide, being NCG (no cover glass). They are intended to be used in metallurgical and epi-illumination applications. Ideal for routine laboratory needs.
Designed to ensure field flatness up to F.N. 22, based on an infinity-corrected optical system.
Documents
Specifikationer
  • Forstørrelse:
    10×/0,25 mm
  • Beskrivelse:
    IOS W-Plan MET objectives (no cover glass)
  • Seq:
    202