VWR®, IOS W-Plan MET Objectives
IOS W-Plan MET objectives (no cover glass), 10×/0,25 mm
Katalog # 630-2161
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These objectives deliver precise performance without the need of the cover slide, being NCG (no cover glass). They are intended to be used in metallurgical and epi-illumination applications. Ideal for routine laboratory needs.
Designed to ensure field flatness up to F.N. 22, based on an infinity-corrected optical system.
Documents
VWR® for Microscopy
Produktlitteratur
Specifikationer
- Forstørrelse:10×/0,25 mm
- Beskrivelse:IOS W-Plan MET objectives (no cover glass)
- Seq:202