Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Desiccator cabinets

Desiccator cabinets

Bench top N₂ purge desiccator cabinet, with static dissipative PVC window, RH% sensor and display panel, 600×400×600 mm
Catalog # PALBUCD-604060
Supplier:   Palbam
Desiccator cabinets
Catalog # PALBUCD-604060
Supplier:  Palbam
Desiccator cabinets
Desiccator cabinets

Bench top N₂ purge desiccator cabinet, with static dissipative PVC window, RH% sensor and display panel, 600×400×600 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Bench top N₂ purge desiccator cabinets made from stainless steel 304, units have an electropolished finish and static dissipative PVC window. Cabinets cut nitrogen expenses by up to 80%, automate clean, dry benchtop storage eliminating moisture & oxygen related degradation and optimise yields. An appropriate flow of nitrogen is introduced into the upper section of the desiccator; the heavier air is then purged out of the bottom. Ideal for use with semiconductor components, biological and pharmaceutical samples, and other sensitive materials. Fully integrated turnkey system takes 30 s to install.

  • Built in RH% sensor and display panel
  • Easy to use control panel allows user to enter the required RH% level
  • Can be used as a single unit on bench top or shelf or as units of two or three in dedicated shelf units
  • N₂ desiccator cabinets minimise the N₂ usage and therefore minimise N₂ leakage into the cleanroom atmosphere

Delivery information: Typically available from stock for direct shipment from the supplier.

Specifications
  • B×D×H udv.:
    600×400×600 mm
  • Beskrivelse:
    Bench top N₂ purge desiccator cabinet, with static dissipative PVC window
  • Seq:
    02
  • med:
    RH% sensor and display panel
Accessories

Product Information

  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • PALBDS8050
    Shelf, solid stainless steel, 800×500 mm
    Desiccator UCD8050
    Log in for availability
    1 * 1 Items

Product Information

  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • PALBDS-6040
    Shelf, solid stainless steel, 600×400 mm
    Desiccator UCD6040
    Log in for availability
    1 * 1 Items

Product Information

  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • PALBDSTAND604040-3
    Desiccator stand, stainless steel, for three units
    Desiccator UCD604040
    Log in for availability
    1 * 1 Items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.