Orders LinkContactUsLinkComponent
Accessories for test sieves
Accessories for test sieves
Sieve pan, 400 mm, 65 mm
Katalog # HAVE205922756
Leverandør:   NEXOPART
Accessories for test sieves
Katalog # HAVE205922756
Leverandør:  NEXOPART
Accessories for test sieves
Accessories for test sieves
Sieve pan, 400 mm, 65 mm
Price
1.460,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Ø:
    400 mm
  • Beskrivelse:
    Sieve pan
  • Højde:
    65 mm
  • Seq:
    11