Orders LinkContactUsLinkComponent
Objectives for PSM-1000 microscopes
Objectives for PSM-1000 microscopes
ELWD plan apo objective (WD = 33,5 mm), 10×/0,28
Katalog # MOTI1101001700032
Leverandør:   MOTIC
Objectives for PSM-1000 microscopes
Katalog # MOTI1101001700032
Leverandør:  MOTIC
Objectives for PSM-1000 microscopes
ELWD plan apo objective (WD = 33,5 mm), 10×/0,28
Price
4.790,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Forstørrelse:
    10×/0,28
  • Beskrivelse:
    ELWD plan apo objective (WD = 33,5 mm)
  • Seq:
    106