Overalls, 4500
hvid, M
Katalog # MMMA4500WM
Leverandør: 3M
Gå til:
Product Details & Documents
This simple overall made of durable PP fabric is suitable for non hazardous applications where hygiene and cleanliness are important.
- Excellent economic solution to keep the wearer clean or dry
- Breathable and light duty protection
- Two-way zip with storm flap
- Elasticated 2-panel hood, cuffs, ankles and waist
Typical applications include: Domestic DIY use, light duty industrial cleaning and maintenance applications.
Weight: 45 g/m²
Certifications: Cat. I
Specifikationer
- Tøj størrelse:M
- Farve:hvid
- Seq:02
- Cat. No.:MMMA4500WM
- Vægt:45 g/m²
- Model:4500
- Outfit applikationer:General Purpose|Laboratory|Industrial
- Stof:PP