Eyepieces
Widefield eyepiece with diamond analyser, 10× (23 mm)
Katalog # MOTI1101001403371
Leverandør: MOTIC
Gå til:
Product Details & Documents
Specifikationer
- Forstørrelse:10× (23 mm)
- Beskrivelse:Widefield eyepiece with diamond analyser
- Seq:103