Caps for sample tubes, tamper evident
Screw cap, with lip seal for tamper evident tubes, 5, 10 and 12 ml, hvid
Katalog # SIMPT552WTP
Leverandør: Simport Scientific
Gå til:
Product Details & Documents
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
- Til rør:5, 10 and 12 ml
- Farve:hvid
- Beskrivelse:Screw cap, with lip seal for tamper evident tubes
- Seq:01