VWR®, N-Plan Objectives
IOS N-plan achromatic objective, 60×/0,80 mm
Katalog # 630-2049
Leverandør: VWR Collection
Gå til:
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
Produktlitteratur
Specifikationer
- Forstørrelse:60×/0,80 mm
- Beskrivelse:IOS N-plan achromatic objective
- Seq:205