Chloroform ≥99.5% stabilised
Glass bottle, 1 L
Supplier: Honeywell Chemicals
Jump to:
Product Details & Documents
Beilstein Number: 1731042
RTECS Number: FS9100000
UNSPSC Code: 12191502
Synonyms: Trichlormethan
- CAS Number:67-66-3
- Formula:CHCl₃
- MDL Number:MFCD00000826
- Molecular Weight:119.38 g/mol
- Boiling Pt:61 °C (1,013 hPa)
- Melting Pt:–61,3 °C
- Melting Pt:-61,3 °C
- Density:1,485 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-663-8
- UN:1888
- ADR:6.1,III
- :stabiliseret med amylen 100-200 ppm
Documents
Specifications
Specification Test Results
- Vapor Pressure:160 mmHg ( 20 °C)
- NMR-spectrum:complying
- Assay (GC):Min. 99.50 %
- 2-methyl-2-buten (GC):20 - 200 ppm
- Refractive Index:n20/D 1.445 (lit.)
Specifications
- Pakning:Glass bottle
- Pakn:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



