Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

SPE columns, Supra-Poly® HLB

HLB, 30 µm, 6 ml, 100 mg
Katalog # PERCN9306672
Leverandør:   PerkinElmer
SPE columns, Supra-Poly® HLB
Katalog # PERCN9306672
Leverandør:  PerkinElmer
SPE columns, Supra-Poly® HLB

HLB, 30 µm, 6 ml, 100 mg

Price
7.010,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
  • Contains macro-porous polymers with ultra-pure, pharmaceutical grade spherical particles
  • Shorter analysis times, greater load capacity and reduced solvent usage
  • Ideal for high throughput assays
Specifikationer
  • Partikelstørrelse:
    30 µm
  • Kapacitet:
    6 ml
  • Seq:
    10
  • Lejevægt:
    100 mg
  • Fase:
    HLB