Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Lotion soaps, Nature

Lotion soap, Nature, bag-in-box, 1,4 l
  PLUM1795
Lotion soaps, Nature
  PLUM1795
Lotion soaps, Nature
Lotion soaps, Nature

Lotion soap, Nature, bag-in-box, 1,4 l

 
101,00 kr
   
1 * 1.400 mL
 

Natural mild, gentle cream soap for normal and frequent hand washing, where special consideration is given to the skin and the environment.

  • With the Swan label
  • Without colourants or perfume
  • For dispenser systems: CombiPlum Electronic (1 L) and MP2000 (1,4 L)

 : *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904

 
  • Beskrivelse:
    Lotion soap, Nature, bag-in-box, 1,4 l
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.