Orders LinkContactUsLinkComponent
Accessories for Midi Plus™ Pipette Controller
Replacement filter, non sterile, 0,45 µm (suitable only for grey toned Midi Plus™)
Katalog # BHITLH-7129120
Leverandør:   Sartorius
Accessories for Midi Plus™ Pipette Controller
Katalog # BHITLH-7129120
Leverandør:  Sartorius
Accessories for Midi Plus™ Pipette Controller
Accessories for Midi Plus™ Pipette Controller
Replacement filter, non sterile, 0,45 µm (suitable only for grey toned Midi Plus™)
Price
1.230,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Replacement filter, non sterile, 0,45 µm (suitable only for grey toned Midi Plus™)
  • Cat. No.:
    BHITLH-7129120
  • Miljøcertificering:
    Renewable energy certificate