Magnetic stirring bars
Magnetic stirring bar, 12 mm, 4,5 mm, ARE/AREX 5 series
Katalog # VELPA00001063
Leverandør: VELP SCIENTIFIC
Gå til:
Product Details & Documents
Specifikationer
- Ø:4,5 mm
- Til:ARE/AREX 5 series
- Beskrivelse:Magnetic stirring bar
- Længde:12 mm
- Seq:02