Magnetic stirring bars
Magnetic stirring bar, 6 mm, 3 mm, ARE/AREX 5 series
Catalog # VELPA00001062
Supplier: VELP SCIENTIFIC
Jump to:
Product Details & Documents
Specifications
- Ø:3 mm
- Til:ARE/AREX 5 series
- Beskrivelse:Magnetic stirring bar
- Længde:6 mm
- Seq:01