Swabs in tubes
Swabs in tube, hvid, Plast / rayon, +
Katalog # GREI425010_1000
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
- Sterilised by irradiation
- Tubes are made of polystyrene
- Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Steril:+
- Beskrivelse:Swabs in tube
- Seq:08
- Skaft/hoved:Plast / rayon
- Lågfarve:hvid