Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, TEMPase Hot Start 2× master mix

VWR®, TEMPase Hot Start 2× master mix

TEMPase Hot Start 2X Master Mix, med Master Mix K, 1,5 mM MgCl₂, 2.500 Tests
Catalog # 733-2582
Supplier:   VWR Chemicals
VWR®, TEMPase Hot Start 2× master mix
Catalog # 733-2582
Supplier:  VWR Chemicals
CAS Number:  
VWR®, TEMPase Hot Start 2× master mix
VWR®, TEMPase Hot Start 2× master mix

TEMPase Hot Start 2X Master Mix, med Master Mix K, 1,5 mM MgCl₂, 2.500 Tests

CAS Number:  
Price
23.840,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

TEMPase Hot Start DNA Polymerase Master Mix and Blue TEMPase Master Mix are good alternatives to TEMPase Hot Start DNA Polymerase. These master mixes offer easy reaction assembly at room temperature, reduced set-up time and fewer handling steps, which lead to increased reproducibility. As a consequence TEMPase Hot Start DNA Polymerase Master Mix is highly suited to standard tests.

The blue loading dye in Blue TEMPase Hot Start DNA Polymerase Master Mix facilitates direct gel loading, and eliminates the need for separate loading dye - no need for time-consuming sample preparation before electrophoresis.

Multiplex 2× Master Mix is composed of TEMPase Hot Start DNA Polymerase and a specialised buffer system designed for multiplex PCR.

Delivery information: TEMPase Hot Start DNA Polymerase Master Mix and Blue TEMPase Master Mix are available in two variations, either based on Key Buffer (Master Mix K) or Combination Buffer (Master Mix C) to suit different PCR requirements. Additional MgCl₂ is included in the kit to enable optimisation.

Specifications

Specifications

  • Pakn:
    2.500 Tests
  • Beskrivelse:
    TEMPase Hot Start 2X Master Mix, med Master Mix K, 1,5 mM MgCl₂
  • Høj troskab:
    No
  • Enheder:
    2500 r Mix 2500
  • Exonuklease aktivitet:
    5’→3’
  • Koncentration:
    2x
  • Buffer sammensætning:
    Based on Ammonium buffer
  • Cat. No.:
    733-2582
  • Størrelse:
    50 x 1.25 ml
  • Beskrivelse:
    TEMPase Hot Start DNA Polymerase 2X Master Mix A<br />1.5 mM MgCl2 2500 reactions
  • Format:
    Mastermix incl. dNTPs
  • Ansøgning:
    Multiplex
  • Amplikon størrelse:
    ≤ 5kb
  • Processivity:
    Standard
  • Product overhang:
    3'A
  • Varmstart:
    Chemically modified
  • Opbevaring betingelser:
    – 20 °C (long term), + 4 °C (6 months)

Specifications