Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Packaging kits, P650/P(I)620

Packaging kits, P650/P(I)620

Packaging set, P650 (UN3373), 500 ml, 100×100×180 mm
Katalog # DAKL520125
Leverandør:   DAKLAPACK
Packaging kits, P650/P(I)620
Katalog # DAKL520125
Leverandør:  DAKLAPACK
Packaging kits, P650/P(I)620

Packaging set, P650 (UN3373), 500 ml, 100×100×180 mm

Price
344,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These kits contain a cardboard outer box (UN3373 or UN2814), a 500 ml green leak proof container, plastic shock absorbers and absorbing tube holders.

  • Non transparent
  • Made from cardboard

Just drop in your samples, seal the lid closed and fit back into the cardboard box and send with your preferred courier.

Specifikationer
  • Kapacitet:
    500 ml
  • Beskrivelse:
    Packaging set, P650 (UN3373)
  • Seq:
    02
  • B×D×H:
    100×100×180 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.