Anti-CTDSPL Rabbit Polyclonal Antibody
25 µL
Katalog # NOVUNB300-231SS
Leverandør: Novus Biologicals
Gå til:
Product Details & Documents
Type: Primary
Antigen: CTDSPL (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Documents
NB300-231.pdf
Datablad
Specifikationer
- Størrelse:25 µL
- Western blot:Yes
- Clonality:Polyclonal
- Gen ID:50511
- Konjugation:Unconjugated
- Antigen synonymer:Synaptonemal complex protein 3|RNASCP3|COR1|Sycp3
- Shipping temperature:Ice
- Antigen name:Synaptonemal Complex Protein 3
- Cat. No.:NOVUNB300-231SS
- Reactivity:Mouse
- Vært:Rabbit
- ImmunoChemistry:Yes
- Antistof type:Primary
- Opbevaringstemperatur:Cool
- Antigen symbol:SCP3
- ImmunFluorescens:Yes



