Orders LinkContactUsLinkComponent
Sterile Serological Pipettes
+, rød, 200 pieces (4×50 pieces), individually blister-wrapped
  EPPE0030127.730
 :   EPPENDORF
Sterile Serological Pipettes
  EPPE0030127.730
 :  EPPENDORF
Sterile Serological Pipettes
Sterile Serological Pipettes
+, rød, 200 pieces (4×50 pieces), individually blister-wrapped
960,00 kr
1 * 200 Items
Eppendorf Serological Pipettes ensure top-tier sterility and safety for sensitive lab applications. They are sterile, free from pyrogens, DNase, RNase, human and bacterial DNA, and non-cytotoxic. Ideal for cell culture and molecular biology tasks.
  • Clear and precise graduations for easy volume determination
  • Color-coded for easy identification of pipet sizes
  • Subunits with dispenser option to keep stored products safe
  • Individually wrapped for reliable sterility
  • Ultra-pure virgin polystyrene meets the requirement of USP VI
  • Harmonized and well-fitting operation with Easypet 3; compatibility with all existing pipet helpers
  • Sterility assurance level of 10⁻⁶
  • Certified free of detectable pyrogens, DNA, RNase, and DNase
  • Certified non-cytotoxic
Designed to work in perfect harmony with the Easypet 3 or Pipet Helper, Eppendorf completes its portfolio in liquid handling with this serological pipette. For ease and flexibility, these serological pipettes can also be used with any other standard pipette controller.
Rely on precise graduations overlaid on ultra-clear pipette material for quick and easy volume readings from 0,1 to 50 ml. Enjoy the freedom of individually wrapped pipettes packaged in a robust dispenser box to ensure consistent premium quality.
  • Farvekode:
    rød
  • Pakket:
    200 pieces (4×50 pieces), individually blister-wrapped
  • Steril:
    +
  • Seq:
    05