Dog serum
Dog serum, Sterile filtered, Frossen
S2900-050
: Biowest
Dog serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Dog serum is used as a blocking agent in immunoassays and controls.
- Serum format:Frossen
- Sterilitet:Sterile filtered
- Beskrivelse:Dog serum
- Seq:20