Orders LinkContactUsLinkComponent
Rabbit plasma
Rabbit plasma with sodium citrate, sterile filtered, 100 ml
Katalog # S2610-100
Leverandør:   Biowest
Rabbit plasma
Katalog # S2610-100
Leverandør:  Biowest
CAS nummer:  
Rabbit plasma
Rabbit plasma with sodium citrate, sterile filtered, 100 ml
CAS nummer:  
Price
320,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.
Specifikationer
Specifikationer
  • Pakn:
    100 ml
  • Beskrivelse:
    Rabbit plasma with sodium citrate, sterile filtered
Specifikationer