Sheep serum
Sheep serum, heat inactivated, Sterile filtered, Frossen
Katalog # S235H-500
Leverandør: Biowest
Gå til:
Product Details & Documents
For immunohistochemistry applications, sheep serum is frequently used as a blocking solution. Using sheep serum is useful for blocking the background cross reactivity with immunoglobulins which are already present in the tissue. When used before application of the primary antibody, sheep serum can eliminate Fc receptor binding of the primary and secondary antibody.
Documents
Specifikationer
- Serum format:Frossen
- Sterilitet:Sterile filtered
- Beskrivelse:Sheep serum, heat inactivated
- Seq:04