Orders LinkContactUsLinkComponent
Trypsin
Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red, 500 ml
Katalog # L0931-500
Leverandør:   Biowest
Trypsin
Katalog # L0931-500
Leverandør:  Biowest
CAS nummer:  
Trypsin
Trypsin
Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red, 500 ml
CAS nummer:  
Price
470,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. The concentration of trypsin necessary to dislodge the cells from their substrate depends on the sensitivity of the cells.
Specifikationer
Specifikationer
  • Pakn:
    500 ml
  • Beskrivelse:
    Trypsin 0,25% EDTA in HBSS, without calcium, without magnesium, with phenol red
Specifikationer