Membrane filters with grid
White with black grid, cellulose nitrate, 6 mm hydrophobic edge, 131, 0,20 µm, 47 mm
Katalog # SART13107-47-HCN
Leverandør: Sartorius
Gå til:
Product Details & Documents
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Ø:47 mm
- Porestørrelse:0,20 µm
- Model:131
- Beskrivelse:White with black grid, cellulose nitrate, 6 mm hydrophobic edge
- Seq:11
- Tykkelse:115 - 145 µm
- Cat. No.:SART13107-47-HCN
- Max. temperature:130 °C